Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Srpr
Entrez Gene ID
67398
Gene Name
signal recognition particle receptor ('docking protein')
Gene Aliases
1300011P19Rik, D11Mgi27
Chromosome Location
Chr.9: 35211203 - 35217003 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026130.1 NP_080406.1 8 - 9 1132 111
GenBank mRNA AK004967.2 - 8 - 9 1132 111
AK038017.1 - 8 - 9 1120 111
AK177061.1 - 3 - 4 262 111
BC012512.1 - - 215 111
BC021839.1 - 8 - 9 1115 111

View Full Product Details