The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037999.2 | NP_001033088.1 | 2 - 3 | 266 | 104 |
NM_007830.4 | NP_031856.1 | 2 - 3 | 260 | 104 | |
GenBank mRNA | AA068307.1 | - | 2 - 3 | 104 | |
AK008540.1 | - | 2 - 3 | 181 | 104 | |
AK008576.1 | - | 2 - 3 | 181 | 104 | |
AK018720.1 | - | 2 - 3 | 182 | 104 | |
AK020692.1 | - | 2 - 3 | 230 | 104 | |
AK027906.1 | - | 2 - 3 | 182 | 104 | |
AK145278.1 | - | 2 - 3 | 182 | 104 | |
AK148753.1 | - | 2 - 3 | 182 | 104 | |
AM000002.1 | - | 2 - 3 | 266 | 104 | |
AV001592.1 | - | 2 - 3 | 154 | 104 | |
BC028874.1 | - | 2 - 3 | 104 | ||
CJ183794.1 | - | 2 - 3 | 104 | ||
EF437365.1 | - | 2 - 3 | 104 | ||
X61431.1 | - | 2 - 3 | 104 |