Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lcn4
Entrez Gene ID
16821
Gene Name
lipocalin 4
Gene Aliases
A630045M08Rik, Vnsp2
Chromosome Location
Chr.2: 26667674 - 26671282 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010695.1 NP_034825.1 5 - 6 518 128
GenBank mRNA BC114544.1 - 5 - 6 517 128
D38581.1 - 5 - 6 518 128

View Full Product Details