Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Srsf4
Entrez Gene ID
57317
Gene Name
serine/arginine-rich splicing factor 4
Gene Aliases
5730499P16Rik, AW550192, MNCb-2616, SRp75, Sfrs4
Chromosome Location
Chr.4: 131873611 - 131901725 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020587.2 NP_065612.2 4 - 5 694 68
GenBank mRNA AB041587.1 - 1 - 2 132 68
AK075769.1 - 4 - 5 662 68
AK077648.1 - 4 - 5 680 68
AK156989.1 - 4 - 5 629 68
BC019437.1 - 4 - 5 654 68
BC026944.1 - 4 - 5 1560 68
BC040812.1 - 4 - 5 1560 68

View Full Product Details