Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mtmr11
Entrez Gene ID
194126
Gene Name
myotubularin related protein 11
Gene Aliases
AI035306, BC051083, Cra
Chromosome Location
Chr.3: 96161946 - 96171718 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181409.3 NP_852074.2 15 - 16 1886 87
GenBank mRNA AK132375.1 - 15 - 16 1853 87
BC042764.1 - 3 - 4 406 87
BC051083.1 - 15 - 16 1819 87

View Full Product Details