Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Taar1
Entrez Gene ID
111174
Gene Name
trace amine-associated receptor 1
Gene Aliases
Tar1, Trar1, taR-1
Chromosome Location
Chr.10: 23920406 - 23921404 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053205.1 NP_444435.1 1 - 1 843 72
GenBank mRNA BC125368.1 - 1 - 1 893 72
BC125370.1 - 1 - 1 893 72

View Full Product Details