Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxl20
Entrez Gene ID
72194
Gene Name
F-box and leucine-rich repeat protein 20
Gene Aliases
2610511F20Rik, 4632423N09Rik, AI849362, AL117906, C86145, Fbl2, Scr, mKIAA4147
Chromosome Location
Chr.11: 98082554 - 98150440 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028149.1 NP_082425.1 15 - 15 8564 151
GenBank mRNA AK028535.1 - 1 - 1 3381 151
BC004087.1 - 1 - 1 1571 151
CX211402.1 - 1 - 1 258 151

View Full Product Details