The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286481.1 | NP_001273410.1 | 5 - 5 | 2241 | 98 |
NM_001286482.1 | NP_001273411.1 | 5 - 5 | 2217 | 98 | |
NM_001286483.1 | NP_001273412.1 | 4 - 4 | 2057 | 98 | |
NM_144841.4 | NP_659090.1 | 3 - 3 | 2139 | 98 | |
GenBank mRNA | AK053741.1 | - | 4 - 4 | 2054 | 98 |