The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175238.5 | NP_780447.4 | 36 - 36 | 8427 | 152 |
GenBank mRNA | AK018316.1 | - | 2 - 2 | 1354 | 152 |
AY585206.1 | - | 35 - 35 | 8420 | 152 | |
BC055320.1 | - | 6 - 6 | 2709 | 152 |