Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gli2
Entrez Gene ID
14633
Gene Name
GLI-Kruppel family member GLI2
Gene Aliases
AW546128
Chromosome Location
Chr.1: 118834061 - 119053619 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081125.1 NP_001074594.1 14 - 14 6653 128
GenBank mRNA AK051858.1 - 1 - 1 2978 128

View Full Product Details