Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gata2
Entrez Gene ID
14461
Gene Name
GATA binding protein 2
Gene Aliases
Gata-2
Chromosome Location
Chr.6: 88192578 - 88207032 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008090.5 NP_032116.4 6 - 6 3035 81
GenBank mRNA AB000096.2 - 6 - 6 2927 81
AK004551.1 - 6 - 6 3032 81
AK154981.1 - 6 - 6 3036 81
AW492264.1 - 1 - 1 81

View Full Product Details