Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gata4
Entrez Gene ID
14463
Gene Name
GATA binding protein 4
Gene Aliases
Gata-4
Chromosome Location
Chr.14: 63198915 - 63271714 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008092.3 NP_032118.2 7 - 7 3311 138
GenBank mRNA AB075549.1 - 7 - 7 3329 138
AK134639.1 - 6 - 6 2335 138
AW558892.1 - 1 - 1 138
CF582373.1 - 1 - 1 138

View Full Product Details