Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Otx1
Entrez Gene ID
18423
Gene Name
orthodenticle homolog 1
Gene Aliases
A730044F23Rik, jv
Chromosome Location
Chr.11: 21994764 - 22001651 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011023.3 NP_035153.1 5 - 5 2756 148
GenBank mRNA AK083671.1 - 5 - 5 2700 148
AK158588.1 - 3 - 3 5114 148
BC058354.1 - 4 - 4 2624 148

View Full Product Details