The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013834.3 | NP_038862.2 | 3 - 3 | 4241 | 126 |
GenBank mRNA | AK008943.1 | - | 1 - 1 | 892 | 126 |
BC094662.1 | - | 3 - 3 | 4227 | 126 |