The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009952.2 | NP_001009952.2 | 1 - 2 | 77 | 80 |
GenBank mRNA | AF272844.1 | - | 1 - 2 | 81 | 80 |
CK616187.1 | - | 1 - 2 | 78 | 80 |