The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021552.5 | NP_067527.3 | 4 - 5 | 692 | 83 |
GenBank mRNA | AF238866.1 | - | 4 - 5 | 618 | 83 |
AK002474.1 | - | 3 - 4 | 543 | 83 | |
AK009136.1 | - | 3 - 4 | 621 | 83 | |
AK012832.1 | - | 4 - 5 | 633 | 83 | |
AK014275.1 | - | 4 - 5 | 669 | 83 | |
AK017605.1 | - | 4 - 5 | 633 | 83 | |
AK078457.1 | - | 4 - 5 | 645 | 83 | |
BC086776.1 | - | 3 - 4 | 488 | 83 | |
BC116384.1 | - | 4 - 5 | 633 | 83 | |
BC126976.1 | - | 4 - 5 | 632 | 83 | |
BC172160.1 | - | 4 - 5 | 604 | 83 |