Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Rplp2
- Entrez Gene ID
-
67186
- Gene Name
- ribosomal protein, large P2
- Gene Aliases
-
2700049I22Rik
- Chromosome Location
-
Chr.7: 141447464 - 141451350 on Build GRCm38
- Gene Symbol
- Rplp2-ps1
- Entrez Gene ID
-
665931
- Gene Name
- ribosomal protein, large P2, pseudogene 1
- Gene Aliases
-
Gm7852
- Chromosome Location
-
Chr.12: 75630918 - 75631749 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_038063.1 |
|
1 - 1 |
429 |
64 |
GenBank mRNA |
BC055860.1 |
- |
1 - 1 |
429 |
64 |
- Gene Symbol
- Wdr89
- Entrez Gene ID
-
72338
- Gene Name
- WD repeat domain 89
- Gene Aliases
-
2600001A11Rik
- Chromosome Location
-
Chr.12: 75630594 - 75669537 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_028203.1 |
NP_082479.1 |
2 - 2 |
2254 |
64 |
GenBank mRNA |
AK132326.1 |
- |
2 - 2 |
2225 |
64 |
View Full Product Details