Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Skint6
Entrez Gene ID
230622
Gene Name
selection and upkeep of intraepithelial T cells 6
Gene Aliases
OTTMUSG00000008519
Chromosome Location
Chr.4: 112804503 - 113286973 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103199.1 NP_001096669.1 4 - 5 758 103
GenBank mRNA EU099304.1 - 4 - 5 758 103
EU099305.1 - 4 - 5 752 103

View Full Product Details