Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vmn2r56
Entrez Gene ID
629079
Gene Name
vomeronasal 2, receptor 56
Gene Aliases
EG629079
Chromosome Location
Chr.7: 12693860 - 12733320 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104648.1 NP_001098118.1 4 - 5 1310 97

View Full Product Details