Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp2j8
Entrez Gene ID
665095
Gene Name
cytochrome P450, family 2, subfamily j, polypeptide 8
Gene Aliases
Cyp2j8-ps, OTTMUSG00000007938
Chromosome Location
Chr.4: 96444283 - 96507386 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104927.1 NP_001098397.1 2 - 3 377 81

View Full Product Details