Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Reck
Entrez Gene ID
53614
Gene Name
reversion-inducing-cysteine-rich protein with kazal motifs
Gene Aliases
St15, mRECK
Chromosome Location
Chr.4: 43875530 - 43944806 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016678.2 NP_057887.2 18 - 19 2537 123
GenBank mRNA AB006960.1 - 18 - 19 2467 123
AK042266.1 - 19 - 20 2716 123
AK143657.1 - 18 - 19 2516 123
AK157885.1 - 18 - 19 2526 123
BC053025.1 - 18 - 19 2476 123
BC132197.1 - 18 - 19 2565 123
BC138065.1 - 18 - 19 2565 123
BC145133.1 - 18 - 19 2566 123

View Full Product Details