The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|---|
RefSeq | NR_002870.2 | 1 - 1 | 6639 | 141 | ||
GenBank mRNA | AK051872.1 | - | LIT3445 D230012O04Rik |
1 - 1 | 2645 | 141 |