The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|---|
RefSeq | NR_003202.1 | 1 - 2 | 227 | 90 | ||
GenBank mRNA | DQ059755.1 | - | LIT3460 PINC |
1 - 2 | 227 | 90 |
DQ059756.1 | - | LIT3461 PINC |
1 - 2 | 227 | 90 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA D530049I02 gene | D530049I02Rik | Anti-sense | Chr. - 73398968 - 73430779 on Build GRCm38 | ||
Downstream | RIKEN cDNA 6030407O03 gene | 6030407O03Rik | 210.8kb | Sense | Chr. - 73618381 - 73864466 on Build GRCm38 |