Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klf14
Entrez Gene ID
619665
Gene Name
Kruppel-like factor 14
Gene Aliases
5330411L03Rik, AL022736, BTEB5
Chromosome Location
Chr.6: 30956021 - 30958990 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135093.1 NP_001128565.1 1 - 1 1153 111
GenBank mRNA AK030435.1 - 2 - 2 447 111
DQ534758.1 - 1 - 1 1145 111

View Full Product Details