Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
Gbp10
Entrez Gene ID
626578
Gene Name
guanylate-binding protein 10
Gene Aliases
-
Chromosome Location
Chr.5: 105215699 - 105239533 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039646.2 NP_001034735.2 1 - 1 55 152
GenBank mRNA BC112328.1 - 1 - 1 72 152
BC114209.1 - 1 - 1 55 152
DQ985743.1 - 1 - 1 55 152

View Full Product Details