Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Vmn2r29
- Entrez Gene ID
-
76229
- Gene Name
- vomeronasal 2, receptor 29
- Gene Aliases
-
6430701C03Rik
- Chromosome Location
-
Chr.7: 7212996 - 7278289 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003555.1 |
|
6 - 7 |
789 |
85 |
GenBank mRNA |
BC096375.1 |
- |
6 - 7 |
715 |
85 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
zinc finger protein 772 |
Zfp772 |
|
-3.0kb
|
Sense |
Chr. - 7202122 - 7209998 on Build GRCm38 |
Downstream |
chloride channel 4-2 |
Clcn4-2 |
Clc4-2, Clcn4 |
47.8kb
|
Sense |
Chr. - 7282309 - 7300851 on Build GRCm38 |
View Full Product Details