Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Dio3os
- Entrez Gene ID
-
353504
- Gene Name
- deiodinase, iodothyronine type III, opposite strand
- Gene Aliases
-
AW556460, Dio3as
- Chromosome Location
-
Chr.12: 110275384 - 110278068 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002866.2 |
|
2 - 3 |
288 |
123 |
GenBank mRNA |
AY283181.1 |
- |
2 - 3 |
288 |
124 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG088109.2 |
- |
2 - 3 |
268 |
|
Chr. 12 - 110276999 - 110278048 [-] on Build GRCm38 |
123 |
GenBankmRNA |
BF469186.1 |
- |
2 - 3 |
404 |
|
Chr. 12 - 110277290 - 110278172 [-] on Build GRCm38 |
123 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
predicted gene, 34785 |
Gm34785 |
|
-93.0kb
|
Sense |
Chr. - 110177956 - 110183644 on Build GRCm38 |
Downstream |
microRNA 1247 |
Mir1247 |
mir-1247, mmu-mir-1247 |
299.0
|
Sense |
Chr. - 110278048 - 110278129 on Build GRCm38 |
View Full Product Details