Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmn2r-ps11
Entrez Gene ID
319210
Gene Name
vomeronasal 2, receptor, pseudogene 11
Gene Aliases
4930518C23Rik
Chromosome Location
Chr.3: 64632863 - 64844743 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_003962.1 8 - 9 2033 60
GenBank mRNA AK029734.1 - 8 - 9 2034 60
AV266491.2 - 3 - 4 449 60


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream vomeronasal 2, receptor 6 Vmn2r6 EG620718, EG667069 -67.4kb Sense Chr. - 64537501 - 64565434 on Build GRCm38
Downstream vomeronasal 2, receptor 7 Vmn2r7 4933425M15Rik 57.1kb Sense Chr. - 64690660 - 64719602 on Build GRCm38

View Full Product Details