Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Snora62
- Entrez Gene ID
-
104433
- Gene Name
- small nucleolar RNA, H/ACA box 62
- Gene Aliases
-
E2, MBI-13, MBI-136, Rnu108
- Chromosome Location
-
Chr.9: 120130434 - 120130563 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002902.2 |
|
1 - 1 |
66 |
105 |
GenBank mRNA |
AF357422.1 |
- |
1 - 1 |
|
105 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
solute carrier family 25, member 38 |
Slc25a38 |
AV019094, BC010801 |
-6.1kb
|
Sense |
Chr. - 120110391 - 120124342 on Build GRCm38 |
Downstream |
predicted gene 5922 |
Gm5922 |
EG546166 |
927.0
|
Anti-sense |
Chr. - 120131490 - 120135371 on Build GRCm38 |
View Full Product Details