The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001139503.1 | NP_001132975.1 | 1 - 2 | 340 | 62 |
GenBank mRNA | BC031595.1 | - | 1 - 2 | 247 | 62 |