The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019461.4 | NP_062334.2 | 1 - 1 | 1606 | 99 |
GenBank mRNA | AF229643.1 | - | 1 - 1 | 898 | 99 |
AK028249.1 | - | - | 1603 | 99 | |
AK030383.1 | - | 1 - 1 | 943 | 99 | |
BC137612.1 | - | 1 - 1 | 1524 | 99 |