The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026751.1 | NP_081027.1 | 1 - 1 | 1153 | 91 |
GenBank mRNA | BC091000.1 | - | 1 - 1 | 926 | 91 |
BC109329.1 | - | 1 - 1 | 892 | 91 | |
BC125008.1 | - | 1 - 1 | 892 | 91 |