Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ttc7
- Entrez Gene ID
-
225049
- Gene Name
- tetratricopeptide repeat domain 7
- Gene Aliases
-
1110035E02Rik, 1700007L07Rik, Ttc7a, fsn, hea
- Chromosome Location
-
Chr.17: 87282082 - 87381770 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK005733.1 |
- |
1 - 2 |
144 |
89 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BY705898.1 |
- |
1 - 2 |
139 |
|
Chr. 17 - 87329789 - 87332049 [-] on Build GRCm38 |
89 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
RIKEN cDNA 4833418N02 gene |
4833418N02Rik |
|
-47.0kb
|
Sense |
Chr. - 87274886 - 87282814 on Build GRCm38 |
Downstream |
RIKEN cDNA 1700011E24 gene |
1700011E24Rik |
GSE |
57.5kb
|
Sense |
Chr. - 87389570 - 87428081 on Build GRCm38 |
View Full Product Details