Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cdh10
- Entrez Gene ID
-
320873
- Gene Name
- cadherin 10
- Gene Aliases
-
A830016G23Rik, C030003B10Rik, C030011H18Rik
- Chromosome Location
-
Chr.15: 18818888 - 19016206 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK021064.1 |
- |
1 - 1 |
946 |
71 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
RIKEN cDNA C030047K22 gene |
C030047K22Rik |
|
-151.0
|
Anti-sense |
Chr. - 18697283 - 18818774 on Build GRCm38 |
Downstream |
predicted gene, 35496 |
Gm35496 |
|
365.3kb
|
Sense |
Chr. - 19185339 - 19203329 on Build GRCm38 |
View Full Product Details