Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Zeb2
- Entrez Gene ID
-
24136
- Gene Name
- zinc finger E-box binding homeobox 2
- Gene Aliases
-
9130203F04Rik, D130016B08Rik, SIP1, Zfhx1b, Zfx1b, Zfxh1b
- Chromosome Location
-
Chr.2: 44983512 - 45113279 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AV375887.2 |
- |
1 |
564 |
|
Chr. 2 - 44983515 - 44984181 [-] on Build GRCm38 |
88 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
glycosyltransferase-like domain containing 1 |
Gtdc1 |
E330008O22Rik |
-55.6kb
|
Sense |
Chr. - 44564412 - 44927918 on Build GRCm38 |
Downstream |
microRNA 5129 |
Mir5129 |
mir-5129, mmu-mir-5129, mu-mir-5129 |
36.9kb
|
Sense |
Chr. - 45023100 - 45023177 on Build GRCm38 |
View Full Product Details