Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Car14
Entrez Gene ID
23831
Gene Name
carbonic anhydrase 14
Gene Aliases
AW536446, Ca14
Chromosome Location
Chr.3: 95897768 - 95905118 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK034693.1 - 1 - 1 1243 72


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB090249.2 - 1 404 Chr. 3 - 95900180 - 95900829 [+] on Build GRCm38 72


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream anterior pharynx defective 1a homolog (C. elegans) Aph1a 6530402N02Rik, APH-1a -744.0 Sense Chr. - 95893907 - 95898592 on Build GRCm38
Downstream acidic (leucine-rich) nuclear phosphoprotein 32 family, member E Anp32e 2810018A15Rik, AI047746, AI326868, CPD1, LANP-L, LANPL, mLANP-L 28.4kb Sense Chr. - 95929250 - 95947387 on Build GRCm38

View Full Product Details