Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Arhgef3
Entrez Gene ID
71704
Gene Name
Rho guanine nucleotide exchange factor (GEF) 3
Gene Aliases
1200004I24Rik, 9830169H03Rik, C76747, Xpln
Chromosome Location
Chr.14: 27114823 - 27403916 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK036737.1 - 1 - 1 454 107


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream interleukin 17 receptor D Il17rd 2810004A10Rik, AI428510, Sef, Sef-S -130.8kb Sense Chr. - 27038801 - 27107286 on Build GRCm38
Downstream family with sequence similarity 208, member A Fam208a 4732418E12, 4933409E02Rik, D14Abb1e 186.2kb Sense Chr. - 27426658 - 27483555 on Build GRCm38

View Full Product Details