Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
A330087I24
Entrez Gene ID
327857
Gene Name
uncharacterized protein A330087I24
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK039689.1 - 2 - 3 421 102


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream vomeronasal 2, receptor 86 Vmn2r86 EG625109 Sense Chr. - 130446199 - 130455894 on Build GRCm38
Downstream vomeronasal 2, receptor 87 Vmn2r87 EG625131 23.1kb Sense Chr. - 130471821 - 130497379 on Build GRCm38

View Full Product Details