Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dennd1b
Entrez Gene ID
329260
Gene Name
DENN/MADD domain containing 1B
Gene Aliases
4632404N19Rik, 4930467M19Rik, 6820401H01Rik, F730008N07Rik
Chromosome Location
Chr.1: 138963446 - 139176043 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166501.1 NP_001159973.1 22 - 22 7549 116
NM_181347.3 NP_851992.1 21 - 21 7415 116
GenBank mRNA AK018353.1 - 1 - 1 1679 116
AK040283.1 - 1 - 1 1646 116


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BM934028.1 - 1 213 Chr. 1 - 139175180 - 139175736 [+] on Build GRCm38 116
GenBankmRNA AA122801.1 - 1 62 Chr. 1 - 139175325 - 139175840 [+] on Build GRCm38 116
GenBankmRNA BG145733.1 - 1 213 Chr. 1 - 139175174 - 139175539 [+] on Build GRCm38 116


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream RIKEN cDNA 4930596I21 gene 4930596I21Rik -194.5kb Sense Chr. - 138977657 - 138979246 on Build GRCm38
Downstream crumbs family member 1, photoreceptor morphogenesis associated Crb1 7530426H14Rik, A930008G09Rik 21.6kb Anti-sense Chr. - 139197546 - 139379118 on Build GRCm38

View Full Product Details