Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hnrnpu
Entrez Gene ID
51810
Gene Name
heterogeneous nuclear ribonucleoprotein U
Gene Aliases
AA408410, AI256620, AL024194, AL024437, AW557595, C86794, Hnrpu, SAFA, Sp120, hnRNP U
Chromosome Location
Chr.1: 178321108 - 178337810 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK049600.1 - 1 - 1 1855 101


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BE951833.1 - 1 Chr. 1 - 178326767 - 178327555 [+] on Build GRCm38 101
GenBankmRNA BX527970.1 - 1 341 Chr. 1 - 178326933 - 178327476 [-] on Build GRCm38 101
GenBankmRNA BB417340.2 - 1 257 Chr. 1 - 178326740 - 178327392 [-] on Build GRCm38 101
GenBankmRNA AV482677.1 - 1 223 Chr. 1 - 178326856 - 178327359 [-] on Build GRCm38 101
GenBankmRNA CN664869.1 - 1 419 Chr. 1 - 178327031 - 178327554 [-] on Build GRCm38 101
GenBankmRNA BF322632.1 - 269 Chr. - - [-] on Build 101
GenBankmRNA AA798850.1 - 1 326 Chr. 1 - 178326912 - 178327461 [-] on Build GRCm38 101
GenBankmRNA BB623648.1 - 1 240 Chr. 1 - 178326733 - 178327375 [-] on Build GRCm38 101


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream COX20 Cox2 chaperone Cox20 2310005N03Rik, Fam36a -4.0kb Anti-sense Chr. - 178319153 - 178322693 on Build GRCm38
Downstream EF-hand calcium binding domain 2 Efcab2 1700073K01Rik, D830011E08Rik 76.9kb Anti-sense Chr. - 178405881 - 178483250 on Build GRCm38

View Full Product Details