Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB456125.2 - 1 575 Chr. 12 - 73038533 - 73039195 [-] on Build GRCm38 100


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream sine oculis-related homeobox 6 Six6 Optx2, Six9 -93.6kb Anti-sense Chr. - 72939945 - 72944899 on Build GRCm38
Downstream sine oculis-related homeobox 1 Six1 BB138287 2.6kb Sense Chr. - 73041827 - 73046712 on Build GRCm38

View Full Product Details