Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Rcor3
- Entrez Gene ID
-
214742
- Gene Name
- REST corepressor 3
- Gene Aliases
-
4921514E24Rik, C730034D20Rik, E130101E15Rik
- Chromosome Location
-
Chr.1: 192098546 - 192138067 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK035958.1 |
- |
1 - 1 |
2915 |
76 |
AK053482.1 |
- |
1 - 1 |
1148 |
76 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BB539863.2 |
- |
1 |
450 |
|
Chr. 1 - 192128108 - 192128750 [-] on Build GRCm38 |
76 |
GenBankmRNA |
BB127325.2 |
- |
1 |
194 |
|
Chr. 1 - 192127850 - 192128536 [-] on Build GRCm38 |
76 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
TNF receptor-associated factor 5 |
Traf5 |
|
-35.5kb
|
Sense |
Chr. - 192052688 - 192092599 on Build GRCm38 |
Downstream |
predicted gene 10516 |
Gm10516 |
ENSMUSG00000073476 |
7.4kb
|
Anti-sense |
Chr. - 192136898 - 192151025 on Build GRCm38 |
View Full Product Details