Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Znrf2
Entrez Gene ID
387524
Gene Name
zinc and ring finger 2
Gene Aliases
1190002C14Rik, AU044205, D6Ertd365e
Chromosome Location
Chr.6: 54816916 - 54890224 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK078666.1 - - 1341 87


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AV317755.2 - 2 455 Chr. 6 - 54890059 - 54890749 [+] on Build GRCm38 87
GenBankmRNA AA790403.1 - 1 124 Chr. 6 - 54890369 - 54890892 [+] on Build GRCm38 87
GenBankmRNA CJ061054.1 - 1 267 Chr. 6 - 54890227 - 54890673 [+] on Build GRCm38 87
GenBankmRNA BE864777.1 - 1 292 Chr. 6 - 54890202 - 54890701 [+] on Build GRCm38 87
GenBankmRNA EH109114.1 - 1 390 Chr. 6 - 54890121 - 54890678 [+] on Build GRCm38 87
GenBankmRNA BU698711.1 - 1 124 Chr. 6 - 54890372 - 54890876 [+] on Build GRCm38 87
GenBankmRNA BB675617.1 - 1 134 Chr. 6 - 54890360 - 54890773 [+] on Build GRCm38 87
GenBankmRNA CX729999.1 - 1 221 Chr. 6 - 54890273 - 54890891 [+] on Build GRCm38 87
GenBankmRNA AA048429.1 - 1 82 Chr. 6 - 54890411 - 54890798 [+] on Build GRCm38 87
GenBankmRNA BB324362.2 - 1 185 Chr. 6 - 54890312 - 54890950 [+] on Build GRCm38 87
GenBankmRNA BU699828.1 - 1 122 Chr. 6 - 54890372 - 54890896 [+] on Build GRCm38 87


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Downstream nucleotide-binding oligomerization domain containing 1 Nod1 C230079P11, Card4, F830007N14Rik, Nlrc1 33.0kb Anti-sense Chr. - 54923942 - 54972672 on Build GRCm38

View Full Product Details