Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nagk
Entrez Gene ID
56174
Gene Name
N-acetylglucosamine kinase
Gene Aliases
Gnk
Chromosome Location
Chr.6: 83795031 - 83802556 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK084646.1 - 1 - 1 1853 109


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA EH103078.1 - 1 551 Chr. 6 - 83796338 - 83796990 [+] on Build GRCm38 109
GenBankmRNA AI481189.1 - 1 Chr. 6 - 83796605 - 83796993 [-] on Build GRCm38 109
GenBankmRNA BB477472.2 - 1 548 Chr. 6 - 83796326 - 83796993 [+] on Build GRCm38 109
GenBankmRNA AI482485.1 - 1 Chr. 6 - 83796633 - 83796999 [-] on Build GRCm38 109
GenBankmRNA BY645188.1 - 1 318 Chr. 6 - 83796556 - 83796992 [+] on Build GRCm38 109
GenBankmRNA BX636794.1 - 1 Chr. 6 - 83796737 - 83796997 [-] on Build GRCm38 109


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream testis expressed gene 261 Tex261 3110001O07Rik, AA409339, AI480706, AL033351, TEG-261 -19.2kb Anti-sense Chr. - 83768881 - 83775814 on Build GRCm38
Downstream poly(A) binding protein interacting protein 2B Paip2b 9.1kb Anti-sense Chr. - 83806058 - 83831741 on Build GRCm38

View Full Product Details