Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Eny2
Entrez Gene ID
223527
Gene Name
enhancer of yellow 2 homolog (Drosophila)
Gene Aliases
1810057B09Rik, 6720481I12, DC6, Ey2
Chromosome Location
Chr.15: 44428081 - 44438256 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK016562.1 - 1 - 1 950 91
AK087816.1 - 1 - 1 952 91


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB556629.2 - 1 413 Chr. 15 - 44428684 - 44429347 [+] on Build GRCm38 91
GenBankmRNA BU518875.1 - 1 503 Chr. 15 - 44428591 - 44429405 [+] on Build GRCm38 91


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream thyrotropin releasing hormone receptor Trhr TRH-R1 -192.2kb Sense Chr. - 44195924 - 44235912 on Build GRCm38
Downstream polycystic kidney and hepatic disease 1-like 1 Pkhd1l1 AB055648, PKHDL1 28.0kb Sense Chr. - 44457371 - 44601376 on Build GRCm38

View Full Product Details