Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Prr14l
- Entrez Gene ID
-
215476
- Gene Name
- proline rich 14-like
- Gene Aliases
-
6030436E02Rik, C330019G07Rik, Prl14l
- Chromosome Location
-
Chr.5: 32789206 - 32854338 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_194340.2 |
NP_919321.2 |
2 - 3 |
254 |
90 |
GenBank mRNA |
AK134346.1 |
- |
2 - 3 |
255 |
90 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CJ049432.1 |
- |
2 - 3 |
255 |
|
Chr. 5 - 32844184 - 32854230 [-] on Build GRCm38 |
90 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
phosphatidylserine decarboxylase |
Pisd |
9030221M09Rik |
-58.4kb
|
Sense |
Chr. - 32736303 - 32785636 on Build GRCm38 |
Downstream |
DEP domain containing 5 |
Depdc5 |
AV016528 |
11.6kb
|
Anti-sense |
Chr. - 32863647 - 32994233 on Build GRCm38 |
View Full Product Details