Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cdc73
- Entrez Gene ID
-
214498
- Gene Name
- cell division cycle 73, Paf1/RNA polymerase II complex component
- Gene Aliases
-
8430414L16Rik, BC027756, C130030P16Rik, C81219, Hrpt2
- Chromosome Location
-
Chr.1: 143607499 - 143702888 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK134896.1 |
- |
- |
1248 |
85 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BE649630.1 |
- |
1 |
355 |
|
Chr. 1 - 143604808 - 143605330 [-] on Build GRCm38 |
85 |
GenBankmRNA |
AW490988.1 |
- |
1 |
|
|
Chr. 1 - 143604725 - 143605175 [+] on Build GRCm38 |
85 |
GenBankmRNA |
BB211070.2 |
- |
1 |
416 |
|
Chr. 1 - 143604735 - 143605387 [-] on Build GRCm38 |
85 |
GenBankmRNA |
BE448747.1 |
- |
1 |
220 |
|
Chr. 1 - 143603719 - 143605195 [-] on Build GRCm38 |
85 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
RIKEN cDNA 4930590L20 gene |
4930590L20Rik |
|
-2.9mb
|
Sense |
Chr. - 140570176 - 140656402 on Build GRCm38 |
View Full Product Details