Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- D730048I06Rik
- Entrez Gene ID
-
68171
- Gene Name
- RIKEN cDNA D730048I06 gene
- Gene Aliases
-
Pate-C, mANLP2
- Chromosome Location
-
Chr.9: 35788050 - 35790112 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK136605.1 |
- |
- |
664 |
91 |
- Gene Symbol
- Gm17727
- Entrez Gene ID
-
100312986
- Gene Name
- predicted gene, 17727
- Gene Aliases
-
Pate-e
- Chromosome Location
-
Chr.9: 35776524 - 35778109 on Build GRCm38
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
predicted gene, 17677 |
Gm17677 |
Pate-n |
-34.3kb
|
Anti-sense |
Chr. - 35741049 - 35742252 on Build GRCm38 |
View Full Product Details