Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ube3a
- Entrez Gene ID
-
22215
- Gene Name
- ubiquitin protein ligase E3A
- Gene Aliases
-
4732496B02, 5830462N02Rik, A130086L21Rik, Hpve6a, mKIAA4216
- Chromosome Location
-
Chr.7: 59227814 - 59311536 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC094605.1 |
- |
1 - 1 |
3357 |
123 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AI154956.1 |
- |
1 |
351 |
|
Chr. 7 - 59309369 - 59309885 [+] on Build GRCm38 |
124 |
GenBankmRNA |
CD552019.1 |
- |
1 |
274 |
|
Chr. 7 - 59309448 - 59309988 [+] on Build GRCm38 |
123 |
GenBankmRNA |
CO045767.1 |
- |
1 |
209 |
|
Chr. 7 - 59309513 - 59310278 [+] on Build GRCm38 |
123 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ATPase, class V, type 10A |
Atp10a |
Atp10c, pfatp |
-479.3kb
|
Sense |
Chr. - 58656143 - 58830035 on Build GRCm38 |
Downstream |
imprinted gene in the Prader-Willi syndrome region |
Ipw |
|
309.3kb
|
Anti-sense |
Chr. - 59619158 - 59678878 on Build GRCm38 |
View Full Product Details