Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Zic1
- Entrez Gene ID
-
22771
- Gene Name
- zinc finger protein of the cerebellum 1
- Gene Aliases
-
ZIC, ZNF201
- Chromosome Location
-
Chr.9: 91358058 - 91365826 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK141431.1 |
- |
1 - 1 |
2050 |
97 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CK780542.1 |
- |
1 |
284 |
|
Chr. 9 - 91358476 - 91359230 [-] on Build GRCm38 |
97 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
predicted gene 28703 |
Gm28703 |
|
-528.3kb
|
Sense |
Chr. - 90824891 - 90829773 on Build GRCm38 |
Downstream |
zinc finger protein of the cerebellum 4 |
Zic4 |
|
783.0
|
Anti-sense |
Chr. - 91361777 - 91389349 on Build GRCm38 |
View Full Product Details